missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring

Mix and store a wide range of culture mediums with these glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

Brand:  Pyrex 1516/09D

Additional Details : Weight : 5.00000kg

 View more versions of this product

Product Code. 15737927

  • € 84.39 / Pack of 10


Estimated Shipment
04-12-2020
Add to basket

Description

Description

  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140°C
Specifications

Specifications

Autoclavable
Yes
Screw Cap
203 mm
750 mL
Ideal for mixing and storage of culture medium
10/Pk.
ISO 4796
Polypropylene
95 mm
Wide
Clear
Borosilicate Glass
Reagent Bottle
Videos
SDS
Documents