Learn More
Pyrex™ Borosilicate Glass Wide Mouth Media Bottles
Manufactured from Pyrex™ borosilicate glass with proven temperature and chemical resistance qualities.
Brand:  Pyrex 1524/12
Additional Details : Weight : 10.35000kg
 View more versions of this product
Product Code. 15747937
  • € 292.00 / Pack of 10
Estimated Shipment
27-11-2020
Add to basket
Description
Description
  • Bottles without screwcap or pouring ring
  • GLS 80™ screwthread provides increased access compared to GL 45 bottles aiding filling and pouring
  • Ideal for use with viscous liquids, pastes, powders, granules and larger solids
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download
  • White enamel graduations and marking spot
  • Autoclavable
  • Screw caps and pouring rings are available separately
Specifications
Specifications
Autoclavable
136 mm
2000 mL
Ideal for use with viscous liquids, pastes, powders, granules and larger solids
10/Pk.
White
No
252 mm
Clear
Borosilicate Glass
Media Bottle