Learn More
Pyrex™ Plain Arms, Straight Bore Stopcock
Click to view available options
Diameter (Metric) Side Arm:
8 mm
9 mm
12.5 mm
Unit Size:
Each
Description
- Glass stopcocks which meet specifications of DIN 12541
- Manufactured from borosilicate glass with interchangeable key
- Leakage rate meets the requirements of DIN 12540 i.e. 0.1mbar l/s-1
- Supplied complete with key retaining device
Specifications
Specifications
| Type | Straight Bore Stopcock |
| Material | Borosilicate Glass |
| Diameter (Metric) Side Arm | 8 mm |
| Certifications/Compliance | DIN 12541 |
| Includes | Key retaining device |