Learn More
Pyrex™ Tall form Berzelius Heavy Duty Beaker
Product Code. p-7116177
Click to view available options
Capacity (Metric):
150 mL
250 mL
600 mL
Unit Size:
Pack of 10
Product Code. 15787917
Brand: Pyrex 1004150
Only null left
Add to basket
Add to basket
Designed for longer life and increased user safety
  • Mechanically stronger than standard beakers
  • Reinforced rims and spout, less prone to breakage during washing etc.
  • Thicker walls and heavier bases
  • Manufactured from Pyrex™ borosilicate glass
  • Printed with Trace Code for downloadable batch certificate
  • White graduations and marking spot
TRUSTED_SUSTAINABILITY
Specifications
Description Beaker
Diameter (Metric) Outer 54mm
Capacity (Metric) 150 mL
Material Borosilicate Glass
Height (Metric) 95mm
Quantity 10/Pk.