Learn More
Pyridine-2-carboxaldoxime, 99%, Thermo Scientific™
Product Code. 11402718
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402718 25 g 25g
11412718 100 g 100g
11422718 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11402718 Supplier Thermo Scientific Alfa Aesar Supplier No. A16714.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 873-69-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006354 MFCD00006354
InChI Key MTFJSAGADRTKCI-VMPITWQZSA-N
Synonym pyridine-2-aldoxime, picolinaldehyde oxime, 2-pyridinecarbaldehyde oxime, z-pyridine-2-carbaldehyde oxime, 2-nitrosomethylidene-1h-pyridine, 2-pyridinealdoxime, e-picolinaldehyde oxime, 2-hydroxyiminomethyl-pyridine, 2-pyridine aldoxime, 2-pyridinecarboxaldehyde, oxime, e
PubChem CID 6740772
IUPAC Name 2-(nitrosomethylidene)-1H-pyridine
SMILES O\N=C\C1=NC=CC=C1
Specifications
Melting Point 110°C to 112°C
Quantity 25 g
Beilstein 109061
Formula Weight 122.13
Percent Purity 99%
Chemical Name or Material Pyridine-2-carboxaldoxime