Learn More
Pyridinium trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
Catalog Number p-7052753
Click to view available options
Quantity:
5 g
25 g
52193-54-1
MFCD00274656
pyridinium trifluoromethanesulfonate, pyridinium triflate, pyridin-1-ium trifluoromethanesulfonate, pyridium triflate, acmc-209ky5, pyridinium trifluoromethanesulphonate, pyridin-1-ium; trifluoromethanesulfonate
52193-54-1
MFCD00274656
pyridinium trifluoromethanesulfonate, pyridinium triflate, pyridin-1-ium trifluoromethanesulfonate, pyridium triflate, acmc-209ky5, pyridinium trifluoromethanesulphonate, pyridin-1-ium; trifluoromethanesulfonate
Pyridinium trifluoromethanesulfonate, esters are used as a catalyst for the reactions of: condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyridinium trifluoromethanesulfonate, esters are used as a catalyst for the reactions of: condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
It is miscible with water, soluble in polar solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 221°C to 223°C
Quantity 5 g
Solubility Information It is miscible with water,soluble in polar solvents.
Formula Weight 229.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Pyridinium trifluoromethanesulfonate