Learn More
Thermo Scientific Chemicals Pyridoxine hydrochloride, 98+%, extra pure
Catalog Number p-3737963
125.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
58-56-0
C8H11NO3·HCl
205.64
MFCD00012807
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
6019
CHEBI:30961
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
CC1=NC=C(C(=C1O)CO)CO.Cl
58-56-0
C8H11NO3·HCl
205.64
MFCD00012807
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
6019
CHEBI:30961
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
CC1=NC=C(C(=C1O)CO)CO.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 214.0°C
Color White
pH 2.4 to 3.0 (5% aq. soln.)
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 50 g
Beilstein 21, V,5, 492
Merck Index 15, 8095
Solubility Information Solution
Formula Weight 205.64
Percent Purity 98+%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Pyridoxine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.