Learn More
Pyronaridine Tetraphosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Pyronaridine Tetraphosphate |
| CAS | 76748-86-2 |
| Molecular Formula | C29H44ClN5O18P4 |
| Quantity | 25 mg |
| Synonym | Benzo[b]-1,5-naphthyridine Phenol Derivative Phosphate, Malaridine Phosphate (1:4), 4-[(7-Chloro-2-methoxybenzo[b]-1,5-naphthyridin-10-yl)amino]-2,6-bis(1-pyrrolidinylmethyl)phenol Phosphate(1:4), 4-((7-Chloro-2-methoxybenzo[b][1,5]naphthyridin-10-yl)amino)-2,6-bis(pyrrolidin-1-ylmethyl)phenol Phosphate(1:4) |
| InChI Formula | InChI=1S/C29H32ClN5O2.4H3O4P/c1-37-26-9-8-24-28(33-26)27(23-7-6-21(30)16-25(23)32-24)31-22-14-19(17-34-10-2-3-11-34)29(36)20(15-22)18-35-12-4-5-13-35;4*1-5(2,3)4/h6-9,14-16,36H,2-5,10-13,17-18H2,1H3,(H,31,32);4*(H3,1,2,3,4) |
| SMILES | COc1ccc2nc3cc(Cl)ccc3c(Nc4cc(CN5CCCC5)c(O)c(CN6CCCC6)c4)c2n1.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O |
| IUPAC Name | 4-[(7-chloro-2-methoxybenzo[b][1,5]naphthyridin-10-yl)amino]-2,6-bis(pyrrolidin-1-ylmethyl)phenol;phosphoric acid |
| Molecular Weight (g/mol) | 910.03 |
| Formula Weight | 909.132 |