missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyronil 45, TRC
Specifications
Specifications
| Chemical Name or Material | Pyronil 45 |
| CAS | 26040-51-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H34 Br4 O4 |
| Quantity | 250 mg |
| Synonym | 1,2-Benzenedicarboxylic acid','3,4,5,6-tetrabromo-','bis(2-ethylhexyl) ester (9CI), Phthalic acid','tetrabromo-','bis(2-ethylhexyl) ester (8CI), Bis(2-ethyl-1-hexyl) tetrabromophthalate, Bis(2-ethylhexyl) 2,3,4,5-Tetrabromophthalate, Bis(2-ethylhexyl) 3,4,5,6-tetrabromophthalate, Bis(2-ethylhexyl) tetrabromophthalate, DP 45, Di(2-ethylhexyl) tetrabromophthalate, FRP 45, Pyronil 45, Uniplex FRP 45 |
| InChI Formula | InChI=1S/C24H34Br4O4/c1-5-9-11-15(7-3)13-31-23(29)17-18(20(26)22(28)21(27)19(17)25)24(30)32-14-16(8-4)12-10-6-2/h15-16H,5-14H2,1-4H3 |
| SMILES | CCCCC(CC)COC(=O)c1c(Br)c(Br)c(Br)c(Br)c1C(=O)OCC(CC)CCCC |
| IUPAC Name | bis(2-ethylhexyl) 3,4,5,6-tetrabromobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 706.14 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?