missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PYROXD1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA558341

Product Code. 15736221

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MCEVKKIYLQ DEFRILKKKS FTFPRDHKSV TADTEMWPVY VELTNEKIYG CDFIVSATGV TPNVEPFLHG NSFDLGEDGG LKVDDHMHTS LPDIYAAG Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.

The PYROXD1 gene encodes the protein pyridine nucleotide-disulphide oxidoreductase domain 1, which has a significant role in mitochondrial function and cellular redox homeostasis. This gene is involved in maintaining the balance of oxidative stress within cells and is crucial for the redox regulation of cellular components, impacting processes like energy metabolism and apoptosis. Mutations in PYROXD1 have been associated with early-onset myopathy, indicating its importance in muscle development and maintenance. The protein encoded by PYROXD1 is part of the oxidoreductase enzyme family that facilitates electron transfer, contributing to cellular defense against oxidative damage. Recent studies have linked PYROXD1 to various neuromuscular disorders, where disruptions in its function affect muscle fiber alignment and force production. Understanding its role in redox biology is crucial for developing therapeutic strategies to combat muscle pathologies and redox-related diseases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PYROXD1
Polyclonal
Unconjugated
PYROXD1
FLJ22028; pyridine nucleotide-disulfide oxidoreductase domain-containing protein 1; pyridine nucleotide-disulphide oxidoreductase domain 1; PYROXD1
Rabbit
Antigen affinity chromatography
RUO
79912
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WU10
PYROXD1
Recombinant protein corresponding to Human PYROXD1. Recombinant protein control fragment (Product #RP-102824).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.