missing translation for 'onlineSavingsMsg'
Learn More

Pyruvate Carboxylase Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA560552

Additional Details : Weight : 0.02000kg

Product Code. 15768381

  • € 530.00 / 100µL
Estimated Shipment: 11-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: GADVVDVAAD SMSGMTSQPS MGALVACTRG TPLDTEVPME RVFDYSEYWE GARGLYAAFD CTATMKSGNS DVYENEIPGG QYTNLHFQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Pyruvate Carboxylase Polyclonal antibody specifically detects Pyruvate Carboxylase in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

Pyruvate Carboxylase
Polyclonal
Unconjugated
PC
EC 6.4.1.1; Pc; Pcb; Pcx; PYC; pyruvate carboxylase; pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; pyruvic carboxylase
Rabbit
Antigen affinity chromatography
RUO
5091
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P11498
PC
Recombinant protein corresponding to Human Pyruvate Carboxylase
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only