Learn More
Pyruvate Kinase Antibody (OTI1H4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44636
Product Code. 30306405
  • € 735.00 / 100µg
Description
Pyruvate Kinase Monoclonal antibody specifically detects Pyruvate Kinase in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pyruvate Kinase
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized
Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence
OTI1H4
Western Blot 1:2000, Flow Cytometry 1:100, Immunocytochemistry/ Immunofluorescence 1:100
PKM, PKM2
Full length human recombinant protein of human PKLR (NP_000289) produced in HEK293T cell.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2a