missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PZP Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5121652

 View more versions of this product

Product Code. 17703054

  • € 208.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PZP Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is highly expressed in late-pregnancy serum and is similar in structure to alpha-2-macroglobulin. The encoded protein, which acts as a homotetramer, inhibits the activity of all four classes of proteinases. This protein contains cleavage sites for several proteinases. Upon binding of a proteinase, the conformation of this protein changes to trap the proteinase, limiting its activity. This protein appears to be elevated in the sera of presymptomatic Alzheimer's disease patients.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PZP
Polyclonal
Unconjugated
PZP
A1m; A1m {ECO:0000312; A2m; AI893533; alpha 1 macroglobulin; alpha-1-M; Alpha-1-macroglobulin; alpha-1-macroglobulin 165 kDa subunit; Alpha-1-macroglobulin 45 kDa subunit; alpha-2-M; alpha-2-macroglobulin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6; CPAMD6; EMBL:AAA40723.1}; MAM; Pregnancy zone protein; pregnancy-zone protein; Pzp; PZP, alpha-2-macroglobulin like
Rabbit
Antigen affinity chromatography, Protein A
RUO
252922
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q63041
PZP
Recombinant protein Pregnancy Zone Protein. The antigen corresponds to amino acid range 1251-1500 of the target protein.
20 μL
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.