missing translation for 'onlineSavingsMsg'
Learn More

Qa-1(b) Mouse anti-Mouse, Brilliant Violet 421, Clone: 6A8.6F10.1A6, BD Optibuild™

Product Code. 15838999
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15838999 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15838999 Supplier BD Biosciences Supplier No. 744385

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 6A8.6F10.1A6 monoclonal antibody reacts with the Qa-1[b] alloantigen, which is a nonclassical MHC class I (Class Ib) molecule encoded by the T23 /gene of the H-2 /complex. Qa-1 associates withβ2-microglobulin and is expressed at low levels on most leukocytes and many other cell types. Its level of cell-surface expression is upregulated by IFNγor specific peptides. Qa-1 is an oligomorphic molecule which presents a limited pool of peptides to T lymphocytes bearingαβandγδTCR and binds to a large subpopulation of NK cells. In fact, Qa-1[b] is the ligand for CD94/NKG2A, CD94/NKG2C, and CD94/NKG2E receptors, which are expressed on NK cells. Furthermore, it has been reported that Qa-1 expressed on activated B lymphocytes is involved in immunoregulation by inducing T-cell-mediated suppression of antibody responses. The 6A8.6F10.1A6 mAb can detect Qa-1[b] on activated splenocytes from C57BL/6 and BALB/c mice (both Qa-1[b]), but not from A/j mice (Qa-1[a]), and it can block target cell recognition by CTLs specific for Qa-1-presented antigen.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Qa-1(b)
Classification Monoclonal
Clone 6A8.6F10.1A6
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Qa-1[b], Qa-1b
Host Species Mouse
Immunogen Qa-1[b] aa. 161-179 Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.