missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QKI Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518576
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, human, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes the sacsin protein, which includes a UbL domain at the N-terminus, a DnaJ domain, and a HEPN domain at the C-terminus. The gene is highly expressed in the central nervous system, also found in skin, skeletal muscles and at low levels in the pancreas. This gene includes a very large exon spanning more than 12.8 kb. Mutations in this gene result in autosomal recessive spastic ataxia of Charlevoix-Saguenay (ARSACS), a neurodegenerative disorder characterized by early-onset cerebellar ataxia with spasticity and peripheral neuropathy. Sacsin has been found to protect against mutant ataxin-1. A pseudogene associated with this gene is located on chromosome 11. Alternative splicing of this gene results in multiple transcript variants.
Specifications
| QKI | |
| Polyclonal | |
| Unconjugated | |
| Qk | |
| 1110003F05Rik; 1500005P18; DKFZp586I0923; HKQ; homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; hqkI; l(17)-1Wis; l17Wis1; lethal, Chr 17, U Wisconsin 1; MqkI; Protein quaking; protein quaking-like; Qk; QK1; QK3; Qka1; Qki; QKI, KH domain containing RNA binding; QKI, KH domain containing, RNA binding; QKI-5 protein; quaking; quaking homolog, KH domain RNA binding; RNA Binding (QKI); RNA binding protein HQK; rqkI | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 19317 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9QYS9 | |
| Qk, QKI | |
| Synthetic peptide sequence (ETKEKPKPTPDY) corresponding to the N-terminus amino acids of QKI (aa 6-18). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction