missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QKI/Quaking Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-239AF532
This item is not returnable.
View return policy
Description
QKI/Quaking Polyclonal antibody specifically detects QKI/Quaking in Human samples. It is validated for Western Blot
Specifications
| QKI/Quaking | |
| Polyclonal | |
| Western Blot | |
| DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
| A synthetic peptide mapping to a region between residues 50 and 100 of isoform 5 of human quaking homolog using the numbering given in entry AAH19917.1 (GeneID 9444). | |
| 0.1 mL | |
| Neuroscience | |
| 9444 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction