missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QKI/Quaking Antibody (S147-6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42196R
This item is not returnable.
View return policy
Description
QKI/Quaking Monoclonal specifically detects QKI/Quaking in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| QKI/Quaking | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| QKI | |
| Fusion protein amino acids 1-341 (full-length) of human QKI-5. Mouse: 100% identity (341/341 amino acids identical). Rat: 99% identity (339/341 amino acids identical) >90% identity with QKI-6, QKI-7 and QKI-7b. | |
| 0.1 mL | |
| Primary | |
| Greater then 90% identity with QKI-6, QKI-7 and QKI-7b.Detects approx 36-38kDa. | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S147-6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
| Mouse | |
| Protein G purified | |
| Neuroscience | |
| 9444 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction