missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QKI/Quaking Antibody (S147-6) - Pan Specific, Novus Biologicals™
Mouse Monoclonal Antibody
€ 174.00 - € 543.00
Specifications
| Antigen | QKI/Quaking |
|---|---|
| Clone | S147-6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605536
|
Novus Biologicals
NBP2-42196-0.025mg |
0.025 mg |
€ 174.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628358
|
Novus Biologicals
NBP2-42196 |
0.1 mg |
€ 543.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
QKI/Quaking Monoclonal antibody specifically detects QKI/Quaking in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| QKI/Quaking | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| PBS (pH 7.4), 50% Glycerol | |
| 9444 | |
| IgG2b | |
| Protein G purified |
| S147-6 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
| Fusion protein amino acids 1-341 (full-length) of human QKI-5. Mouse: 100% identity (341/341 amino acids identical). Rat: 99% identity (339/341 amino acids identical) >90% identity with QKI-6, QKI-7 and QKI-7b. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title