missing translation for 'onlineSavingsMsg'
Learn More

QNZ46, MedChemExpress

Product Code. 16432837
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432837

Brand: Medchem Express HY15703

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress QNZ46 is a NR2C/NR2D-selective NMDA receptor non-competitive antagonist (IC50 values are 3, 6, 229, and >300, >300 μM for NR2D, NR2C, NR2A, NR2B, and GluR1, respectively).

QNZ46 is a NR2C/NR2D-selective NMDA receptor non-competitive antagonist (IC50 values are 3, 6, 229, and >300, >300 μM for NR2D, NR2C, NR2A, NR2B, and GluR1, respectively).

Specifications

Chemical Name or Material QNZ46
CAS 1237744-13-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H17N3O6
Quantity 50 mg
Solubility Information DMSO : ≥ 4.5 mg/mL (10.15 mM)
SMILES O=C(O)C1=CC=C(N2C(/C=C/C3=CC=CC([N+]([O-])=O)=C3)=NC4=C(C=C(OC)C=C4)C2=O)C=C1
Molecular Weight (g/mol) 443.41
Formula Weight 443.41
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.