missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QPRT Monoclonal Antibody (OTI1C10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525200
This item is not returnable.
View return policy
Description
QPRT Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
QPRT gene encodes a key enzyme in catabolism of quinolinate, an intermediate in the tryptophan-nicotinamide adenine dinucleotide pathway. Quinolinate acts as a most potent endogenous exitotoxin to neurons. Elevation of quinolinate levels in the brain has been linked to the pathogenesis of neurodegenerative disorders such as epilepsy, Alzheimer's disease, and Huntington's disease. Alternative splicing results in multiple transcript variants.
Specifications
| QPRT | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15274 | |
| QPRT | |
| Full length human recombinant protein of QPRT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1C10 | |
| Unconjugated | |
| QPRT | |
| 2410027J01Rik; AI647766; epididymis secretory sperm binding protein Li 90n; HEL-S-90n; nicotinate-nucleotide pyrophosphorylase; nicotinate-nucleotide pyrophosphorylase (carboxylating); nicotinate-nucleotide pyrophosphorylase [carboxylating]; QAPRTase; QPRT; QPRTase; quinolinate phosphoribosyltransferase; Quinolinate phosphoribosyltransferase [decarboxylating] | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23475 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction