missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QRSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548981
This item is not returnable.
View return policy
Description
Predicted to react with monkey based on sequence homology.
Allows the formation of correctly charged Gln-tRNA(Gln) through the transamidation of misacylated Glu-tRNA(Gln) in the mitochondria. The reaction takes place in the presence of glutamine and ATP through an activated gamma-phospho-Glu- tRNA(Gln).
Specifications
| QRSL1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H0R6 | |
| QRSL1 | |
| KLH conjugated synthetic peptide between 129-156 amino acids from the N-terminal region of human QRSL1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| QRSL1 | |
| 2700038P16Rik; C80053; GatA; Glu-AdT subunit A; glu-AdT subunit A {ECO:0000255; glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1; glutaminyl-tRNA synthase-like protein 1; glutaminyl-tRNA synthase-like protein 1 {ECO:0000255; glutamyl-tRNA(Gln) amidotransferase subunit A homolog; glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial {ECO:0000255; glutamyl-tRNA(Gln) amidotransferase, subunit A; HAMAP-Rule:MF_03150}; QRSL1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 55278 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction