missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QSOX1/Quiescin Q6 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81797AF532
This item is not returnable.
View return policy
Description
QSOX1/Quiescin Q6 Polyclonal antibody specifically detects QSOX1/Quiescin Q6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| QSOX1/Quiescin Q6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| EC 1.8.3.2, FLJ34858, hQSOX, Q6, QSCN6, Quiescin Q6, quiescin Q6 sulfhydryl oxidase 1, sulfhydryl oxidase 1 | |
| QSOX1 antibody was raised against a 17 amino acid peptide near the center of human QSOX1. The immunogen is located within amino acids 360 - 410 of QSOX1. Amino Acid Squence: FKTALDDRKEGAVLAKK | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5768 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction