missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QSOX1/Quiescin Q6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33198
This item is not returnable.
View return policy
Description
QSOX1/Quiescin Q6 Polyclonal specifically detects QSOX1/Quiescin Q6 in Human samples. It is validated for Western Blot.
Specifications
| QSOX1/Quiescin Q6 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| QSOX1 | |
| Recombinant protein encompassing a sequence within the center region of human QSOX1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.3mg/mL | |
| Western Blot 1:500-1:3000 | |
| EC 1.8.3.2, FLJ34858, hQSOX, Q6, QSCN6, Quiescin Q6, quiescin Q6 sulfhydryl oxidase 1, sulfhydryl oxidase 1 | |
| Rabbit | |
| 83 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5768 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction