missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QTRT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15759771 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15759771 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15759771 Supplier Invitrogen™ Supplier No. PA561970

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LRKKVFEKDF GPIDPECTCP TCQKHSRAFL HALLHSDNTA ALHHLTVHNI AYQLQLMSAV RTSIVEKRFP DFVRDFMGAM YGDPTLCPTW A Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 76%.

tRNA-guanine transglycosylase (TGT; EC 2.4.2.29) synthesizes queuosine (Q), which is found in tRNAs that recognize NAU and NAC codons, encoding tyr, asn, asp, and his. Prokaryotic TGT is a single protein of 43 kD. In contrast, mammalian TGT appears to be a heterodimer consisting of a 60-kD subunit (USP14; MIM 607274) and a 43-kD catalytic subunit (QTRT1) (Deshpande and Katze, 2001 [PubMed 11255023]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen QTRT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene QTRT1
Gene Accession No. Q9BXR0
Gene Alias 2610028E17Rik; FP3235; Guanine insertion enzyme; QTRT1; queuine tRNA-ribosyltransferase; queuine tRNA-ribosyltransferase 1; queuine tRNA-ribosyltransferase 1 (tRNA-guanine transglycosylase); queuine tRNA-ribosyltransferase catalytic subunit 1; queuine tRNA-ribosyltransferase; queuine tRNA-ribosyltransferase catalytic subunit 1; similar to queuine tRNA-ribosyltransferase 1; Tgt; TGT, 43-KD subunit; TGT, catalytic subunit; TGUT; tRNA-guanine transglycosylase; zgc:66378
Gene Symbols QTRT1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human QTRT1 (Product #RP-99236)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.