missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QTRTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557838
This item is not returnable.
View return policy
Description
Immunogen sequence: DKPRLISGVS RPDEVLECIE RGVDLFESFF PYQVTERGCA LTFSFDYQPN PEETLLQQNG TQEEIKCMDQ IKKIETTGC Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
Interacts with QTRT1 to form an active queuine tRNA-ribosyltransferase. This enzyme exchanges queuine for the guanine at the wobble position of tRNAs with GU(N) anticodons (tRNA-Asp, -Asn, -His and -Tyr), thereby forming the hypermodified nucleoside queuosine (Q) (7-(((4,5-cis-dihydroxy-2-cyclopenten-1-yl)amino)methyl)-7-deazaguanosine).
Specifications
| QTRTD1 | |
| Polyclonal | |
| Unconjugated | |
| QTRT2 | |
| 3110012M05Rik; 4930470H18Rik; AI648807; FLJ12960 protein; Qtrt2; Qtrtd1; Queuine tRNA-ribosyltransferase accessory subunit 2; queuine tRNA-ribosyltransferase domain containing 1; queuine tRNA-ribosyltransferase domain-containing protein 1; queuine tRNA-ribosyltransferase subunit QTRTD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H974 | |
| QTRT2 | |
| Recombinant protein corresponding to Human QTRTD1. Recombinant protein control fragment (Product #RP-96375). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction