Learn More
Julabo™ Quad Distributing Adapter
Quad distributing adapter with barbed fittings
Brand:  Julabo™ 8970522
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11924768
  • € 1125.00 / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Quad distributing adapter with barbed fittings
Specifications
Specifications
19.05mm Tubing
Quad Distributing Adapter
19.05 mm