Learn More
Julabo™ Quad Distributing Adapter
Click to view available options
Connection Type:
1x M24 x 1.5 F to 4x M24 x 1.5 M
1x M30 x 1.5 F to 4x M30 x 1.5 M
1x M38 x 1.5 F to 4x M38 x 1.5 M
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System |
| Type | Quad Distributing Adapter |
| Connection Type | 1x M24 x 1.5 F to 4x M24 x 1.5 M |