Learn More
Julabo™ Quad Distributing Adapter
Product Code. 15274303
Click to view available options
Connection Type:
1x M24 x 1.5 F to 4x M24 x 1.5 M
1x M30 x 1.5 F to 4x M30 x 1.5 M
1x M38 x 1.5 F to 4x M38 x 1.5 M
Unit Size:
Each
Product Code. 15274303
Brand: Julabo™ 8890141
Only null left
Specifications
For Use With (Equipment) Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System
Type Quad Distributing Adapter
Connection Type 1x M24 x 1.5 F to 4x M24 x 1.5 M