Learn More
Honeywell™ QUAD (i)XTREM S3 Shoes
Honeywell™ QUAD (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
€ 178.00 - € 188.00
Specifications
Certifications/Compliance | EN ISO 20345:2011 S3 HRO HI CI SRC |
---|---|
Color | Brown, Black |
Material | Leather |
Sole Material | Polyurethane and nitrile |
Toe Style | Metal-free |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15856595
|
Honeywell™
6246212-37/7 |
37 |
€ 178.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15866595
|
Honeywell™
6246212-38/7 |
38 |
€ 178.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15876595
|
Honeywell™
6246212-39/7 |
39 |
€ 178.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15886595
|
Honeywell™
6246212-40/7 |
40 |
€ 188.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15896595
|
Honeywell™
6246212-41/7 |
41 |
€ 178.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15806605
|
Honeywell™
6246212-42/7 |
42 |
€ 188.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15816605
|
Honeywell™
6246212-43/7 |
43 |
€ 188.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15826605
|
Honeywell™
6246212-44/7 |
44 |
€ 178.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15836605
|
Honeywell™
6246212-45/7 |
45 |
€ 188.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15846605
|
Honeywell™
6246212-46/7 |
46 |
€ 188.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15856605
|
Honeywell™
6246212-47/7 |
47 |
€ 188.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15866605
|
Honeywell™
6246212-48/7 |
48 |
€ 178.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15876605
|
Honeywell™
6246212-49/7 |
49 |
€ 178.00
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Descripción
- Water resistant grain full leather
- Comfort foam for improved foot stability
- Gusset to avoid water and dust penetration
- Poromax 3D lining
- Toe cap metal free
- Metal free antiperforation insert
- Injected PU / Nitrile sole
- (i)NTELLIGENT wear control
- EN ISO 20345:2011 S3 HRO HI CI SRC
Especificaciones
EN ISO 20345:2011 S3 HRO HI CI SRC | |
Leather | |
Metal-free |
Brown, Black | |
Polyurethane and nitrile |