Learn More
ValidR BV Qualification NEN EN 12469
Product Code. 30041489
Click to view available options
For Use With (Equipment):
Labconco PSM and BSC Cabinets
Unit Size:
Each
Product Code. 30041489
Brand: ValidR BV BSC_NEN_EN_12469_5
Qualification (or Validation) of Biosafety Cabinets of norm NEN EN 12469.
  • The EN 12469 certification is a European standard that establishes the performance requirements for biosafety cabinets concerning microbiological safety.
  • EN 12469:2000 specifies the basic requirements for microbiological safety cabinets (MSCs) concerning safety and hygiene.
  • This standard sets the minimum performance criteria for safety cabinets for work with micro-organisms.
  • It specifies test procedures for microbiological safety cabinets concerning protecting both the worker and the environment, product protection and cross-contamination.
Specifications
For Use With (Equipment) Labconco PSM and BSC Cabinets
Product Type Performance Check