Learn More
ValidR BV Qualification NEN EN 12469
Click to view available options
For Use With (Equipment):
Labconco PSM and BSC Cabinets
Unit Size:
Each
Description
- The EN 12469 certification is a European standard that establishes the performance requirements for biosafety cabinets concerning microbiological safety.
- EN 12469:2000 specifies the basic requirements for microbiological safety cabinets (MSCs) concerning safety and hygiene.
- This standard sets the minimum performance criteria for safety cabinets for work with micro-organisms.
- It specifies test procedures for microbiological safety cabinets concerning protecting both the worker and the environment, product protection and cross-contamination.
Specifications
Specifications
| For Use With (Equipment) | Labconco PSM and BSC Cabinets |
| Product Type | Performance Check |