missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Qualitative Water Finder Test Papers

Qualitative Water Finder Test Paper allows the quick and easy detection of water and water vapor, such as the control of leak tightness of tubing, gas streams, and much more.

Brand:  Macherey-Nagel™ 90630

Additional Details : Weight : 0.01000kg

Product Code. 16249061

  • € 61.30 / 7m
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • For water and vapor detection
  • 7 m length reel
  • 2 year shelf life
Specifications

Specifications

White to Pink/Lavender (Water)
Detection of Water and Vapor
7 m
Reel
Test Paper
14 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers