Learn More
Socorex™ Qualitix™ Micropipette Tips, 10μL
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 10μL also come in a variety of shapes to facilitate fluid transfer.
Brand:  Socorex™ 3090.0010RS
Additional Details : Weight : 0.63000kg
 View more versions of this product
Product Code. 15750669
  • € 84.39 / Pack of 1152
Estimated Shipment
02-12-2020
Add to basket
Description
Description
  • Cover a range of sizes.
  • Autoclavable.
  • Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
  • Various tip versions available.
Specifications
Specifications
10μL
Sterile
Autoclavable
Micropipette
96/Rack
Non-filtered
Natural
Yes
Ultra-micro Tip, Fine Point
12/PK, 10PK/CS