Learn More
Socorex™ Qualitix™ Micropipette Tips, 1000μL, natural
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 1000μL also come in a variety of shapes to facilitate fluid transfer.
€ 40.70 - € 86.40
Specifications
Volume (Metric) | 1000 μL |
---|---|
Volume (Metric) Dispensing | 1000μL |
Color | Natural |
Autoclavable | Autoclavable |
Disposable | Single-use |
Product Code | Brand | Packaging | Sterility | Tip Style | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Sterility | Tip Style | Quantity | Price | Quantity & Availability | ||
15730659
|
Socorex™
3070.1000R |
96/Rack | Non-sterile | Fine Point | 6/PK, 8PK/CS |
€ 75.80
Pack of 576 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
15740659
|
Socorex™
3070.1000RS |
96/Rack | Sterile | Fine Point | 6/PK, 8PK/CS |
€ 86.40
Pack of 576 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||
15760669
|
Socorex™
3090.1000B |
1000/Bag | Non-sterile | Regular Tip, Bevel Point | 1/PK 10PK/CS |
€ 40.70
Pack of 1000 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||
15770669
|
Socorex™
3090.1000R |
96/Rack | Non-sterile | Regular Tip, Bevel Point | 6/PK, 8PK/CS |
€ 75.80
Pack of 576 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||
15780649
|
Socorex™
3070.1000B |
1000/Bag | Non-sterile | Fine Point | 1/PK, 10PK/CS |
€ 40.70
Pack of 1000 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||
Description
- Cover a range of sizes.
- Autoclavable.
- Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
- Various tip versions available.
Specifications
1000 μL | |
Natural | |
Single-use |
1000μL | |
Autoclavable | |
Qualitix |