Learn More
Socorex™ Qualitix™ Micropipette Tips, 1000μL, natural
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 1000μL also come in a variety of shapes to facilitate fluid transfer.
€ 40.70 - € 86.40
Specifications
Volume (Metric) 1000 μL
Volume (Metric) Dispensing 1000μL
Color Natural
Autoclavable Autoclavable
Disposable Single-use
View More Specs
Products 5
Product Code Brand Packaging Sterility Tip Style Quantity Price Quantity & Availability  
Product Code Brand Packaging Sterility Tip Style Quantity Price Quantity & Availability  
15730659
View Documents Promotion Details
Socorex™
3070.1000R
96/Rack Non-sterile Fine Point 6/PK, 8PK/CS
€ 75.80
Pack of 576
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
15740659
View Documents Promotion Details
Socorex™
3070.1000RS
96/Rack Sterile Fine Point 6/PK, 8PK/CS
€ 86.40
Pack of 576
View alternative products
Save Up to 
»
Estimated Shipment: 09-07-2024
to see stock.
Add to basket
15760669
View Documents Promotion Details
Socorex™
3090.1000B
1000/Bag Non-sterile Regular Tip, Bevel Point 1/PK 10PK/CS
€ 40.70
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
15770669
View Documents Promotion Details
Socorex™
3090.1000R
96/Rack Non-sterile Regular Tip, Bevel Point 6/PK, 8PK/CS
€ 75.80
Pack of 576
View alternative products
Save Up to 
»
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
15780649
View Documents Promotion Details
Socorex™
3070.1000B
1000/Bag Non-sterile Fine Point 1/PK, 10PK/CS
€ 40.70
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Cover a range of sizes.
  • Autoclavable.
  • Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
  • Various tip versions available.
Specifications
Specifications
1000 μL
Natural
Single-use
1000μL
Autoclavable
Qualitix