missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Qualitix™ Micropipette Tips, 200μL

Product Code. 15760679
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
1000/Bag
96/Rack
Tip Style:
Beveled
Gel Loading, Capillary
Regular Tip, Bevel Point
Unit Size:
Pack of 1000
Pack of 576
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Packaging Tip Style unitSize
15760679 Non-sterile 1000/Bag Beveled Pack of 1000
15700649 Non-sterile 1000/Bag Beveled Pack of 1000
15740649 Non-sterile 96/Rack Beveled Pack of 960
15750649 Sterile 96/Rack Beveled Pack of 960
15770679 Non-sterile 96/Rack Beveled Pack of 960
15780659 Non-sterile 96/Rack Gel Loading, Capillary Pack of 576
15780679 Sterile 96/Rack Regular Tip, Bevel Point Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15760679 Supplier Socorex™ Supplier No. 3290.0200B
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 200μL also come in a variety of shapes to facilitate fluid transfer.

  • Cover a range of sizes.
  • Autoclavable.
  • Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
  • Various tip versions available.

Specifications

Volume (Metric) 200 μL
Volume (Metric) Dispensing 200μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Tip Style Beveled
Packaging 1000/Bag
Quantity 1/PK 10PK/CS
Disposable Single-use
For Use With (Equipment) Socorex
Product Line Qualitix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.