missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Qualitix™ Micropipette Tips, 200μL

Product Code. 15770679
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
1000/Bag
96/Rack
Tip Style:
Beveled
Gel Loading, Capillary
Regular Tip, Bevel Point
Unit Size:
Pack of 1000
Pack of 576
Pack of 960
This item is not returnable. View return policy

Product Code. 15770679

Brand: Socorex™ 3290.0200R

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 200μL also come in a variety of shapes to facilitate fluid transfer.

  • Cover a range of sizes.
  • Autoclavable.
  • Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
  • Various tip versions available.

Specifications

Volume (Metric) 200 μL
Volume (Metric) Dispensing 200μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Tip Style Beveled
Packaging 96/Rack
Quantity 10/PK, 10PK/CS
Disposable Single-use
For Use With (Equipment) Socorex
Product Line Qualitix
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.