Learn More
Socorex™ Qualitix™ Micropipette Tips, 200μL
Description
- Cover a range of sizes.
- Autoclavable.
- Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
- Various tip versions available.
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Volume (Metric) Dispensing | 200μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Natural |
| Autoclavable | Autoclavable |
| Tip Style | Beveled |
| Packaging | 1000/Bag |
| Quantity | 1/PK, 10PK/CS |
| Disposable | Single-use |
| Show More |