Learn More
Socorex™ Qualitix™ Micropipette Tips, 200μL
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 200μL also come in a variety of shapes to facilitate fluid transfer.
€ 29.60 - € 110.00
Specifications
Volume (Metric) 200 μL
Volume (Metric) Dispensing 200μL
Filtered Non-Filtered
Autoclavable Autoclavable
Disposable Single-use
View More Specs
Products 7
Product Code Brand Packaging Sterility Tip Style Price Quantity & Availability  
Product Code Brand Packaging Sterility Tip Style Price Quantity & Availability  
15700649
View Documents Promotion Details
Socorex™
3070.0200B
1000/Bag Non-sterile Universal, Bevel Point
€ 29.60
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
15740649
View Documents Promotion Details
Socorex™
3070.0200R
96/Rack Non-sterile Universal, Bevel Point
€ 69.80
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 01-05-2024
to see stock.
Add to basket
15750649
View Documents Promotion Details
Socorex™
3070.0200RS
96/Rack Sterile Universal, Bevel Point
€ 79.30
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
15760679
View Documents Promotion Details
Socorex™
3290.0200B
1000/Bag Non-sterile Regular Tip, Bevel Point
€ 29.60
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
15770679
View Documents Promotion Details
Socorex™
3290.0200R
96/Rack Non-sterile Regular Tip, Bevel Point
€ 69.80
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
15780659
View Documents Promotion Details
Socorex™
3080.0200GR
96/Rack Non-sterile Gel Loading, Capillary
€ 110.00
Pack of 576
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
15780679
View Documents Promotion Details
Socorex™
3290.0200RS
96/Rack Sterile Regular Tip, Bevel Point
€ 79.30
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
Add to basket
Description
Description
  • Cover a range of sizes.
  • Autoclavable.
  • Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
  • Various tip versions available.
Specifications
Specifications
200 μL
Non-Filtered
Single-use
Qualitix
200μL
Autoclavable
Socorex™