Learn More
Hellma™ Quartz Suprasil Fluorescence Cells
Brand:  Hellma™ 221001-10-40
Additional Details : Weight : 0.50000kg
Product Code. 10773317
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Fluorescence Cell
Quartz Suprasil
1Each
200 to 2500 nm
12.5 mm
3.5 mL
10mm
10 x 10 mm
40 mm