Learn More
Hellma™ Quartz Suprasil Fluorescence Cells
Product Code. 13312310
Click to view available options
Height (Metric):
40 mm
45 mm
49.5 mm
56 mm
Unit Size:
Each
Pack of 2
Product Code. 13312310
Brand: Hellma™ 2211040
Only null left
Specifications
Type Fluorescence Cell
Capacity (Metric) 3500 μL
Material Quartz Suprasil
Width (Metric) Interior 10mm
Quantity 1 Each
Path Length 10 x 10 mm
Height (Metric) 40 mm
Width (Metric) 12.5 mm