Learn More
Hellma™ Quartz Suprasil Ultra-Micro Absorption Cells
Product Code. p-8009480
Click to view available options
Capacity (Metric):
0.7 to 5 μL
100 μL
300 μL
Unit Size:
Each
Pack of 2
Product Code. 10648972
Brand: Hellma™ 1052011540
Only null left
Specifications
Type Ultra-micro Absorption Cell
Capacity (Metric) 0.7 to 5 μL
Material Quartz Suprasil
Quantity 1 Each
Path Length 10 mm
Height (Metric) 45 mm
For Use With (Equipment) Photometers and Spectrophotometers
Width (Metric) 12.5 mm