missing translation for 'onlineSavingsMsg'
Learn More

Quassin, TRC

Product Code. 30011421
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011421 2.5 mg 2.50mg
30005636 1 mg 1mg
30007565 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011421 Supplier Toronto Research Chemicals Supplier No. TRCQ5094032.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-78-8
Molecular Formula C22 H28 O6
Molecular Weight (g/mol) 388.4541
Synonym Picrasa-2,12-diene-1,11,16-trione, 2,12-dimethoxy-, Phenanthro[10,1-bc]pyran-1,5,11(4H)-trione, 3aβ,6aβ,7,7aα,8,11a,11bα,11c-octahydro-2,10-dimethoxy-3,8α,11aβ,11cβ-tetramethyl- (8CI), Quassin (6CI,7CI), Phenanthro[10,1-bc]pyran, picrasa-2,12-diene-1,11,16-trione deriv., (+)-Quassin, 2,12-Dimethoxypicrasa-2,12-diene-1,11,16-trione, NSC 36342, Nigakilactone D, Quassine
SMILES COC1=C[C@@H](C)[C@@H]2C[C@H]3OC(=O)C[C@H]4C(=C(OC)C(=O)[C@@H]([C@@]34C)[C@@]2(C)C1=O)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C22H28O6/c1-10-7-14(26-5)20(25)22(4)12(10)8-15-21(3)13(9-16(23)28-15)11(2)18(27-6)17(24)19(21)22/h7,10,12-13,15,19H,8-9H2,1-6H3/t10-,12+,13+,15-,19+,21-,22+/m1/s1
Formula Weight 388.1886
Percent Purity >95%
Chemical Name or Material Quassin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.