Learn More
Bel-Art™ Quaver™ and Quaverette™ Vibrating Spatulas
Product Code. 11307184 Shop All SP Scienceware Products
Click to view available options
Product Type:
Quaver™ Vibrating Spatula
Quaverette™ Micro-Blade Vibrating Spatula
Blade Material:
Stainless Steel
Stainless Steel, Fluo-Kem™ Teflon™ FEP Coating
Shape:
Flat, Curved End
Narrow
Length (English):
7 in.
8.25 in.
Unit Size:
Each
Product Code. 11307184
Brand: Bel-Art™ H367150000
Only null left
Add to basket
Add to basket
Accurately transfer small amounts of material
  • Rotating the toothed thumb wheel vibrates the spatula, giving user control over transfer from spatula to weighing dish
  • Make accurate additions of precious materials as needed
  • Quaver™ spatula features a flat blade with curved end
Specifications
Autoclavable Not Autoclavable
Shape Flat, Curved End
Coating Brown Fluo-Kem™ Teflon™ FEP
Certifications/Compliance FDA
Product Type Quaver™ Vibrating Spatula
For Use With (Equipment) Weighing dish or weighing boat or container
For Use With (Application) Accurately transfer small amounts of material, provides precise control of sample transfer from the blade to the weighing dish or weighing boat
Length (English) 7 in.
Blade Material Stainless Steel, Fluo-Kem™ Teflon™ FEP Coating
Includes Angled end, toothed thumb wheel
Material Stainless Steel
Length (Metric) 18 cm