missing translation for 'onlineSavingsMsg'
Learn More

Quercetin, TRC

Product Code. 30010219
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010219 25 g 25g
30008221 5 g 5g
18010740 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010219 Supplier Toronto Research Chemicals Supplier No. TRCQ51003825G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-39-5
Molecular Formula C15 H10 O7
Molecular Weight (g/mol) 302.24
Synonym 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-, Flavone, 3,3',4',5,7-pentahydroxy- (7CI,8CI), Flavone, 3,4',5,5',7-pentahydroxy- (6CI), 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-benzopyran-4-one, 3,3',4',5,7-Pentahydroxyflavone, 3,5,7,3',4'-Pentahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4-dihydroxyphenyl)-4H-chromen-4-one, 3'-Hydroxykaempferol, C.I. 75670, Corvitin, Cyanidelonon 1522, Korvitin, LDN 0052529, Lipoflavon, Meletin, NSC 57655, NSC 9219, Quercetin, Quercetine, Quercetol, Quertin, Quertine, Sophoretin, Xanthaurine
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one
SMILES OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H10O7/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6/h1-5,16-19,21H
Formula Weight 302.0427
Percent Purity >95%
Chemical Name or Material Quercetin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.