Learn More
Quetiapine Sulfone, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Quetiapine Sulfone
Specifications
Specifications
| Chemical Name or Material | Quetiapine Sulfone |
| CAS | 329216-65-1 |
| Analyte or Component Names | Quetiapine Sulfone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H25 N3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-[2-[4-(5,5-Dioxidodibenzo[b,f][1,4]thiazepin-11-yl)-1-piperazinyl]ethoxy]ethanol, Quetiapine Sulfone |
| InChI Formula | InChI=1S/C21H25N3O4S/c25-14-16-28-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)29(26,27)20-8-4-2-6-18(20)22-21/h1-8,25H,9-16H2 |
| SMILES | OCCOCCN1CCN(CC1)C2=Nc3ccccc3S(=O)(=O)c4ccccc24 |
| IUPAC Name | 2-[2-[4-(11,11-dioxobenzo[b][1,4]benzothiazepin-6-yl)piperazin-1-yl]ethoxy]ethanol |
| Show More |