missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Quick Charger

Product Code. 11761086
Click to view available options
Color:
Black
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11761086

Brand: Brady™ 710615

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Quick Charger for BMP71 printers.

  • Accessory for BMP71 printer.

Specifications

Product Type AC Adapter
Color Black
For Use With (Equipment) BMP71 Label Printer
Description Quick Charger, UK 240 V
Format AC Adapter
Quantity 1 Ea.
Voltage 240 V
Alternate Part Number M71-QC-UK
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.