Learn More
Quickfit™ Borosilicate Glass Centrifuge Tubes
Click to view available options
Capacity (Metric):
10 mL
30 mL
Diameter (Metric):
17 mm
28.5 mm
Thread Size:
14/15
24/20
Unit Size:
Pack of 10
Description
- Borosilicate glass with precision ground Quickfit™socket to accept tapered stopper
- Conical or cylindrical shape
- Each tube is marked with its weight to the nearest 0.1g to enable tubes of similar weight to be selected from any batch
- Can be centrifuged at RCF values up to 3, 000
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Material | Borosilicate Glass |
| Thread Size | 24/20 |
| Shape | Cylindrical |
| Length (Metric) | 115 mm |
| Diameter (Metric) | 28.5 mm |
| Max. RCF | 3000 x g |