Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Product Code. 15557800
Click to view available options
For Use With (Equipment):
19/26 socket size
34/35 socket size
40/38 socket size
Unit Size:
Pack of 5
Product Code. 15557800
Brand: Quickfit EX13/12
Designed to support Raschig rings within plain fractionating columns.
  • Support is passed through socket of column and rests at the constriction above the cone
  • Manufactured from chemically resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Designed to support Raschig rings within plain fractionating columns
Type Column packing support
Material Pyrex Borosilicate Glass
For Use With (Equipment) 19/26 socket size