Learn More
Quickfit™ Chromatography Columns with Rotaflo™ Stopcock and Ground Cone
Manufactured from borosilicate glass. Quickfit™ Chromatography Columns with Rotaflo™ Stopcock and Ground Cone includes glass sintered disc (porosity 0) to support column packing. GP Rotaflo™ stopcock with PTFE key provides fine control of eluent flow. Stem within lower cone allows mounting on a Büchner flask for vacuum flash chromatography.
Brand:  Quickfit CR12/30
Additional Details : Weight : 0.11000kg
Product Code. 12011272
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • With glass sintered disc (porosity 0) to support column packing
  • GP Rotaflo™ stopcock with PTFE key for fine control of eluent flow
  • Stem within lower cone allows mounting on a Büchner flask for vacuum flash chromatography
  • Manufactured from borosilicate glass
Specifications
Specifications
10 mm
1 Pack
Pyrex™ Borosilicate Glass
GP Rotaflo™ stopcock with PTFE key, Glass sintered disc
Chromatography Column
470 mm
19/26 Ground cone