Learn More
Quickfit™ Glass Stirrer Gland
Product Code. 10238024
Click to view available options
Standard Taper Joint:
19/26
24/29
29/32
Unit Size:
Each
Pack of 5
Product Code. 10238024
Brand: Quickfit ST20/2
Only null left
Add to basket
Add to basket
Introduce stirrer paddles into Quickfit™ flasks and vessels with these glass stirrer glands. Quickfit™ Glass Stirrer Glands provide a vapor-tight seal between paddle and flask.
  • Holds a vacuum down to 0.1 torr (13.3Pa)
  • Comprises parallel ground glass sleeve with cup to accept a suitable lubricant
  • Supplied complete with screwcaps rubber rings and PTFE washer
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Quickfit flasks and vessels
For Use With (Application) For introducing stirrer paddles into Quickfit flasks and vessels
Includes Screw cap, Rubber rings and PTFE washers
Standard Taper Joint 19/26