Learn More
Quickfit™ Pig Receiver
Click to view available options
Cone Size:
14/23
Unit Size:
Each
Description
- Allows up to three flasks* to be connected to a condenser outlet
- By slightly rotating the adapter a new flask is presented to the distillate
- With sidearm for connection to a vacuum supply
Specifications
Specifications
| Cone Size | 14/23 |
| For Use With (Equipment) | Flask |
| Height (English) | 4.33in. |
| Height (Metric) | 110mm |
| Includes | Side arm |
| Material | Borosilicate Glass |
| Product Type | Bend adapter |